missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylbismuth Difluoride 97.0+%, TCI America™
$333.05 - $1122.38
Chemical Identifiers
| CAS | 2023-48-5 |
|---|---|
| Molecular Formula | C18H15BiF2 |
| Molecular Weight (g/mol) | 478.295 |
| InChI Key | OSGJIYNQSIPLAQ-UHFFFAOYSA-L |
| Synonym | Difluorotriphenylbismuth |
| PubChem CID | 4573913 |
| IUPAC Name | difluoro(triphenyl)bismuth |
| SMILES | C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F |
Chemical Identifiers
| 2023-48-5 | |
| 478.295 | |
| Difluorotriphenylbismuth | |
| difluoro(triphenyl)bismuth |
| C18H15BiF2 | |
| OSGJIYNQSIPLAQ-UHFFFAOYSA-L | |
| 4573913 | |
| C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F |
Specifications
| 2023-48-5 | |
| White-Yellow | |
| 1 g | |
| OSGJIYNQSIPLAQ-UHFFFAOYSA-L | |
| difluoro(triphenyl)bismuth | |
| 4573913 | |
| ≥97.0% (T) | |
| Triphenylbismuth Difluoride |
| 159°C | |
| C18H15BiF2 | |
| Difluorotriphenylbismuth | |
| C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F | |
| 478.295 | |
| 478.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator