missing translation for 'onlineSavingsMsg'
Learn More

Triphenylbismuth Difluoride 97.0+%, TCI America™

Catalog No. T26571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26571G 1 g
T26575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26571G Supplier TCI America Supplier No. T26571G

Chemical Identifiers

CAS 2023-48-5
Molecular Formula C18H15BiF2
Molecular Weight (g/mol) 478.295
InChI Key OSGJIYNQSIPLAQ-UHFFFAOYSA-L
Synonym Difluorotriphenylbismuth
PubChem CID 4573913
IUPAC Name difluoro(triphenyl)bismuth
SMILES C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 478.30
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylbismuth Difluoride
TSCA No
Recommended Storage Refrigerator