missing translation for 'onlineSavingsMsg'
Learn More

Triphenylamine, 99+%

Catalog No. p-3758110
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140310250 25 g Glass bottle
AC140311000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140310250 Supplier Thermo Scientific Chemicals Supplier No. 140310250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 603-34-9 | C18H15N | 245.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 603-34-9
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.32
MDL Number MFCD00003020
InChI Key ODHXBMXNKOYIBV-UHFFFAOYSA-N
Synonym triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2
PubChem CID 11775
IUPAC Name N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 125°C to 127°C
Color White to Beige
Boiling Point 347°C to 348°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (C6H5)3N
Quantity 25 g
Beilstein 12, 181
Solubility Information Solubility in water: insoluble
Formula Weight 245.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Triphenylamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-035-5

RUO – Research Use Only