missing translation for 'onlineSavingsMsg'
Learn More

Triphenylamine, 98%

Catalog No. AAA1593922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1593922 100 g
AAA1593906 5 g
AAA1593914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1593922 Supplier Thermo Scientific Chemicals Supplier No. A1593922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 603-34-9 | C18H15N | 245.325 g/mol

Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 603-34-9
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD00003020
InChI Key ODHXBMXNKOYIBV-UHFFFAOYSA-N
Synonym triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2
PubChem CID 11775
IUPAC Name N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 125°C to 129°C
Boiling Point 365°C
Quantity 100 g
Beilstein 2050487
Solubility Information Insoluble in water.
Formula Weight 245.33
Percent Purity 98%
Chemical Name or Material Triphenylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-035-5
RTECSNumber YK2680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only