missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylacetic Acid 97.0+%, TCI America™
$242.33 - $969.30
Chemical Identifiers
| CAS | 595-91-5 |
|---|---|
| Molecular Formula | C20H16O2 |
| Molecular Weight (g/mol) | 288.35 |
| MDL Number | MFCD00004185 |
| InChI Key | DCYGAPKNVCQNOE-UHFFFAOYSA-N |
| Synonym | triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927 |
| PubChem CID | 68992 |
| IUPAC Name | triphenylacetic acid |
| SMILES | OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 595-91-5 | |
| 288.35 | |
| DCYGAPKNVCQNOE-UHFFFAOYSA-N | |
| 68992 | |
| OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H16O2 | |
| MFCD00004185 | |
| triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927 | |
| triphenylacetic acid |
Specifications
| 595-91-5 | |
| C20H16O2 | |
| 5 g | |
| DCYGAPKNVCQNOE-UHFFFAOYSA-N | |
| triphenylacetic acid | |
| 68992 | |
| ≥97.0% (T) | |
| Triphenylacetic Acid |
| 270°C | |
| MFCD00004185 | |
| triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927 | |
| OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 288.35 | |
| 288.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : No