missing translation for 'onlineSavingsMsg'
Learn More

Triphenylacetic Acid 97.0+%, TCI America™

Catalog No. T15855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15855G 5 g
T158525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15855G Supplier TCI America Supplier No. T15855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 595-91-5
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004185
InChI Key DCYGAPKNVCQNOE-UHFFFAOYSA-N
Synonym triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
PubChem CID 68992
IUPAC Name triphenylacetic acid
SMILES OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 270°C
Quantity 5 g
Formula Weight 288.35
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylacetic Acid