missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Phosphite 97.0+%, TCI America™

Catalog No. T0510500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0510500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0510500G Supplier TCI America Supplier No. T0510500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 101-02-0
Color Yellow
Boiling Point 360°C
Molecular Formula C18H15O3P
Quantity 500 g
MDL Number MFCD00003032
UN Number 3082
Synonym phosphorous acid, triphenyl ester, advance tpp, triphenoxyphosphine, phenyl phosphite, stabilizer p 36, trifenoxyfosfin, trifenylfosfit, phosclere t 36, efed, mellite 310
InChI Key HVLLSGMXQDNUAL-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3
IUPAC Name triphenyl phosphite
Molecular Weight (g/mol) 310.289
PubChem CID 7540
Formula Weight 310.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triphenyl Phosphite
Show More Show Less
EINECSNumber (3)-2501
RTECSNumber TH1575000
TSCA Yes