missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphite, 97%

Catalog No. p-7024284
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1866236 500 g
AAA186620E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1866236 Supplier Thermo Scientific Chemicals Supplier No. A1866236
Only null left

CAS: 101-02-0 | C18H15O3P | 310.289 g/mol

Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.

Solubility
Insoluble in water.

Notes
Air and Moisture Sensitive, store away form air and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

Chemical Identifiers

CAS 101-02-0
Molecular Formula C18H15O3P
Molecular Weight (g/mol) 310.289
MDL Number MFCD00003032
InChI Key HVLLSGMXQDNUAL-UHFFFAOYSA-N
Synonym phosphorous acid, triphenyl ester, advance tpp, triphenoxyphosphine, phenyl phosphite, stabilizer p 36, trifenoxyfosfin, trifenylfosfit, phosclere t 36, efed, mellite 310
PubChem CID 7540
IUPAC Name triphenyl phosphite
SMILES C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3

Specifications

Melting Point 22°C to 24°C
Density 1.184
Boiling Point 360°C
Flash Point 146°C (295°F)
Odor Mild
Linear Formula (C6H5O)3P
Quantity 500 g
UN Number UN3077
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 310.29
Percent Purity 97%
Chemical Name or Material Triphenyl phosphite
Show More Show Less
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-908-4
RTECSNumber TH1575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only