missing translation for 'onlineSavingsMsg'
Learn More

Tripalmitin 80.0+%, TCI America™

Catalog No. p-7137573
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T309625G 25 g
T3096100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T309625G Supplier TCI America Supplier No. T309625G
Only null left

Chemical Identifiers

CAS 555-44-2
Molecular Formula C51H98O6
Molecular Weight (g/mol) 807.339
MDL Number MFCD00008995
InChI Key PVNIQBQSYATKKL-UHFFFAOYSA-N
Synonym tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
PubChem CID 11147
ChEBI CHEBI:77393
IUPAC Name 2,3-di(hexadecanoyloxy)propyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 320°C
Quantity 25 g
Formula Weight 807.34
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tripalmitin
EINECSNumber (2)-0669
RTECSNumber RT4953500
TSCA Yes