accessibility menu, dialog, popup

UserName

H3COOOOCH3OOCH3

CAS RN 555-44-2

IUPAC Name: 1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Synonyms: tripalmitin palmitin, tri- tripalmitylglycerol glycerin tripalmitate spezialfett 116 palmitic triglyceride glycerol tripalmitate tripalmitin (substance) palmitic acid triglycerin ester barolub lcd
Molecular Weight (g/mol): 807.34
Molecular Formula: C51H98O6
InChi Key: PVNIQBQSYATKKL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (3)
  • (7)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)