missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tripalmitin | |
| 66°C | |
| 320°C | |
| MFCD00008995 | |
| tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate | |
| CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC | |
| 807.339 | |
| CHEBI:77393 | |
| ≥85.0% (GC) |
| 555-44-2 | |
| White-Yellow | |
| C51H98O6 | |
| 250 g | |
| PVNIQBQSYATKKL-UHFFFAOYSA-N | |
| 2,3-di(hexadecanoyloxy)propyl hexadecanoate | |
| 11147 | |
| 807.34 | |
| Crystalline Powder |
Chemical Identifiers
| 555-44-2 | |
| 807.339 | |
| PVNIQBQSYATKKL-UHFFFAOYSA-N | |
| 11147 | |
| 2,3-di(hexadecanoyloxy)propyl hexadecanoate |
| C51H98O6 | |
| MFCD00008995 | |
| tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate | |
| CHEBI:77393 | |
| CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0669
RTECSNumber : RT4953500
TSCA : Yes