missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triolein, ≥95%, MP Biomedicals™
Triolein is used as a substrate for microbial lipases. Triolein is a symmetrical triglyceride
$178.24 - $178.24
Chemical Identifiers
| CAS | 122-32-7 |
|---|---|
| Molecular Formula | C57H104O6 |
| Molecular Weight (g/mol) | 885.453 |
| MDL Number | MFCD00137563 |
| InChI Key | PHYFQTYBJUILEZ-BSCDBXJPSA-N |
| Synonym | triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate |
| PubChem CID | 45253964 |
| IUPAC Name | [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
| SMILES | CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10312205
|
MP Biomedicals Inc
0210312205 |
5 g |
Each for $178.24
|
|
|||||
Chemical Identifiers
| 122-32-7 | |
| 885.453 | |
| PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
| 45253964 | |
| CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
| C57H104O6 | |
| MFCD00137563 | |
| triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
| [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
Specifications
| 122-32-7 | |
| 0.91g/mL(lit.) | |
| C57H104O6 | |
| 5 g | |
| PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
| [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate | |
| 45253964 | |
| Liquid |
| Yellow | |
| ≥95% | |
| MFCD00137563 | |
| triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
| CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC | |
| 885.453 | |
| 885.46 | |
| Triolein |
Safety and Handling
EINECSNumber : 204-534-7
Recommended Storage : Store at +4°C.