missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylvinylammonium Bromide 97.0+%, TCI America™
Supplier: TCI America T049410G
Specifications
| Trimethylvinylammonium Bromide | |
| White-Yellow | |
| MFCD00050204 | |
| Neurine Bromide | |
| C[N+](C)(C)C=C.Br | |
| 167.07 | |
| 166.06 | |
| Crystalline Powder |
| 10603-92-6 | |
| C5H13BrN+ | |
| 10 g | |
| IDVSELVVGYIOEX-UHFFFAOYSA-N | |
| ethenyl(trimethyl)azanium;hydrobromide | |
| 57345485 | |
| ≥97.0% (T) |
Chemical Identifiers
| 10603-92-6 | |
| 167.07 | |
| IDVSELVVGYIOEX-UHFFFAOYSA-N | |
| 57345485 | |
| C[N+](C)(C)C=C.Br |
| C5H13BrN+ | |
| MFCD00050204 | |
| Neurine Bromide | |
| ethenyl(trimethyl)azanium;hydrobromide |
Safety and Handling
RTECSNumber : BT3675000
TSCA : No