missing translation for 'onlineSavingsMsg'
Learn More

Trimethylvinylammonium Bromide 97.0+%, TCI America™

Catalog No. p-7136312
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04941G 1 g
T049410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04941G Supplier TCI America Supplier No. T04941G
Only null left

Chemical Identifiers

CAS 10603-92-6
Molecular Formula C5H13BrN+
Molecular Weight (g/mol) 167.07
MDL Number MFCD00050204
InChI Key IDVSELVVGYIOEX-UHFFFAOYSA-N
Synonym Neurine Bromide
PubChem CID 57345485
IUPAC Name ethenyl(trimethyl)azanium;hydrobromide
SMILES C[N+](C)(C)C=C.Br

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 166.06
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylvinylammonium Bromide
RTECSNumber BT3675000
TSCA No