missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin chloride, 99%

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol

$575.68 - $575.68

Chemical Identifiers

CAS 1066-45-1
Molecular Formula C3H11ClSn
Molecular Weight (g/mol) 201.28
MDL Number MFCD00000520
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
PubChem CID 14016
IUPAC Name chloro(trimethyl)stannane
SMILES Cl.C[SnH](C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC163990500
SDS View Documents
Thermo Scientific Chemicals
163990500
50 g Glass bottle
Each for $575.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1066-45-1
201.28
FVFGWISLDLUGHX-UHFFFAOYSA-N
14016
Cl.C[SnH](C)C
C3H11ClSn
MFCD00000520
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
chloro(trimethyl)stannane

Specifications

1066-45-1
White
97°C
98.5% min. (GC)
C3H11ClSn
MFCD00000520
17,388; 03,19; 04,27; 05,26; 06,41; 07,18; 13,239
Solubility in water: soluble. Other solubilities: soluble in chloroform,organic solvents
Cl.C[SnH](C)C
201.28
199.25
Crystals
37.0°C to 39.0°C
148.0°C
Authentic
Glass bottle
(CH3)3SnCl
50 g
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
FVFGWISLDLUGHX-UHFFFAOYSA-N
chloro(trimethyl)stannane
14016
99%
Trimethyltin chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 213-917-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only