accessibility menu, dialog, popup

UserName

CAS RN 1066-45-1

IUPAC Name: trimethyltin chloride
Synonyms: trimethyltin chloride
Molecular Weight (g/mol): 201.28
Molecular Formula: C3H11ClSn
InChi Key: FVFGWISLDLUGHX-UHFFFAOYSA-N
SMILES: Cl.C[SnH](C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)