missing translation for 'onlineSavingsMsg'
Learn More

Trimethylthiourea 98.0+%, TCI America™

Catalog No. T0667100G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0667100G 100 g
T0667500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0667100G Supplier TCI America Supplier No. T0667100G
Only null left

Chemical Identifiers

CAS 2489-77-2
Molecular Formula C4H10N2S
Molecular Weight (g/mol) 118.198
MDL Number MFCD00059148
InChI Key JAEZSIYNWDWMMN-UHFFFAOYSA-N
PubChem CID 2779938
IUPAC Name 1,1,3-trimethylthiourea
SMILES CNC(=S)N(C)C

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 100 g
Formula Weight 118.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylthiourea
EINECSNumber (2)-1766
RTECSNumber YU4900000
TSCA Yes