missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium chloride, 97%

Catalog No. p-3790850
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC381520010 1 g Glass bottle
AC381520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC381520010 Supplier Thermo Scientific Chemicals Supplier No. 381520010

CAS: 5034-06-0 | C3H9ClOS | 128.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 5034-06-0
Molecular Formula C3H9ClOS
Molecular Weight (g/mol) 128.62
MDL Number MFCD00011898
InChI Key KQYWHJICYXXDSQ-UHFFFAOYSA-M
Synonym trimethylsulfoxonium chloride, trimethyloxosulfonium chloride, trimethylsulfonium chloride oxide, trimethyloxosulphonium chloride, sulfonium, trimethyl-, chloride, s-oxide, sulfoxonium, trimethyl-, chloride, sulfoxonium, trimethyl-, chloride 8ci,9ci, trimethyl oxo-??-sulfanylium chloride, c3h9os.cl, sulfoxonium, chloride
PubChem CID 197819
SMILES C[S+](=O)(C)C.[Cl-]

Specifications

Melting Point 226.0°C to 229.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3S(O)Cl
Quantity 1 g
Formula Weight 128.62
Percent Purity 97%
Chemical Name or Material Trimethylsulfoxonium chloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-724-6

RUO – Research Use Only