accessibility menu, dialog, popup

UserName

ClCH3SCH3CH3O

CAS RN 5034-06-0

IUPAC Name: trimethyl(oxo)-λ⁶-sulfanylium chloride
Synonyms: trimethyl(oxo)-λ⁶-sulfanylium chloride
Molecular Weight (g/mol): 128.61
Molecular Formula: C3H9ClOS
InChi Key: KQYWHJICYXXDSQ-UHFFFAOYSA-M
SMILES: [Cl-].C[S+](C)(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results