missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsulfonium hydroxide, 0.25M solution in methanol
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Trimethylsulfonium hydroxide |
| Color | Colorless |
| Name Note | 0.25M solution in methanol |
| CAS | 17287-03-5, 67-56-1 |
| Molecular Formula | C3H9IS |
| MDL Number | MFCD00011632 |
| Concentration | 0.2 to 0.3M |
| Density | 0.81g/mL |
| Synonym | trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent |
| InChI Key | VFJYIHQDILEQNR-UHFFFAOYSA-M |
| Show More |
RUO – Research Use Only