missing translation for 'onlineSavingsMsg'
Learn More

Trimethylstearylammonium Chloride 98.0+%, TCI America™

Catalog No. p-7135801
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S008725G 25 g
S0087100G 100 g
S0087500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S008725G Supplier TCI America Supplier No. S008725G

Chemical Identifiers

CAS 112-03-8
Molecular Formula C21H46ClN
Molecular Weight (g/mol) 348.06
MDL Number MFCD00050188
InChI Key VBIIFPGSPJYLRR-UHFFFAOYSA-M
Synonym n,n,n-trimethyloctadecan-1-aminium chloride, stac, octadecyltrimethylammonium chloride, cation ab, stearyltrimethylammonium chloride, steartrimonium chloride, trimethyloctadecylammonium chloride, 1-octadecanaminium, n,n,n-trimethyl-, chloride, nissan cation ab, aliquat 7
PubChem CID 8155
ChEBI CHEBI:81710
IUPAC Name trimethyl(octadecyl)azanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 348.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylstearylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BT1750000
TSCA Yes