missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilylmethylmagnesium Chloride (ca. 18% in Tetrahydrofuran, ca. 1mol/L), TCI America™
Supplier: TCI America T2609100ML
Specifications
| Trimethylsilylmethylmagnesium Chloride (ca. 18% in Tetrahydrofuran, ca. 1mol/L) | |
| C4H11ClMgSi | |
| 100 mL | |
| trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride | |
| C[Si](C)(C)C[Mg]Cl | |
| 146.97 | |
| 146.97 |
| 13170-43-9 | |
| MFCD00009916 | |
| 3399 | |
| NAQATMJWCJCHOZ-UHFFFAOYSA-M | |
| [(chloromagnesio)methyl]trimethylsilane | |
| 2734900 |
Chemical Identifiers
| 13170-43-9 | |
| 146.97 | |
| NAQATMJWCJCHOZ-UHFFFAOYSA-M | |
| 2734900 | |
| C[Si](C)(C)C[Mg]Cl |
| C4H11ClMgSi | |
| MFCD00009916 | |
| trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride | |
| [(chloromagnesio)methyl]trimethylsilane |
Safety and Handling
TSCA : No