missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylmethyl Acetate 98.0+%, TCI America™

Catalog No. A16135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16135G 5 g
A161325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16135G Supplier TCI America Supplier No. A16135G
Only null left

Chemical Identifiers

CAS 2917-65-9
Molecular Formula C6H14O2Si
Molecular Weight (g/mol) 146.26
MDL Number MFCD00009650
InChI Key TXZMVBJSLYBOMN-UHFFFAOYSA-N
Synonym trimethylsilyl methyl acetate, acetoxymethyltrimethylsilane, methanol, trimethylsilyl-, acetate, methanol, 1-trimethylsilyl-, 1-acetate, 2-trimethylsilyl methyl acetate, acmc-1ci3n, o-, acetoxymethyl trimethyl silane, txzmvbjslybomn-uhfffaoysa, acetic acid trimethylsilylmethyl ester
PubChem CID 76214
IUPAC Name (trimethylsilyl)methyl acetate
SMILES CC(=O)OC[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
UN Number 3272
Formula Weight 146.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylmethyl Acetate