CAS RN 2917-65-9
IUPAC Name:
(trimethylsilyl)methyl acetate
Synonyms:
(trimethylsilyl)methyl acetate
Molecular Weight (g/mol):
146.26
Molecular Formula:
C6H14O2Si
InChi Key:
TXZMVBJSLYBOMN-UHFFFAOYSA-N
SMILES:
CC(=O)OC[Si](C)(C)C
1
–
1
of
1
results
1
Trimethylsilylmethyl Acetate 98.0+%, TCI America™
CAS: 2917-65-9 Molecular Formula: C6H14O2Si Molecular Weight (g/mol): 146.26 MDL Number: MFCD00009650 InChI Key: TXZMVBJSLYBOMN-UHFFFAOYSA-N Synonym: trimethylsilyl methyl acetate,acetoxymethyltrimethylsilane,methanol, trimethylsilyl-, acetate,methanol, 1-trimethylsilyl-, 1-acetate,2-trimethylsilyl methyl acetate,acmc-1ci3n,o-,acetoxymethyl trimethyl silane,txzmvbjslybomn-uhfffaoysa,acetic acid trimethylsilylmethyl ester PubChem CID: 76214 IUPAC Name: (trimethylsilyl)methyl acetate SMILES: CC(=O)OC[Si](C)(C)C