missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylmethanol 97.0+%, TCI America™

Supplier:  TCI America T12505ML

 View more versions of this product

Catalog No. T12505ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethylsilylmethanol
Yellow
C4H12OSi
5 mL
trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrr
C[Si](C)(C)CO
104.224
104.22
Liquid
3219-63-4
121°C
MFCD00004596
1993
ZQKNBDOVPOZPLY-UHFFFAOYSA-N
trimethylsilylmethanol
76692
≥97.0% (GC)

Chemical Identifiers

3219-63-4
104.224
ZQKNBDOVPOZPLY-UHFFFAOYSA-N
76692
C[Si](C)(C)CO
C4H12OSi
MFCD00004596
trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrr
trimethylsilylmethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : PC4100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions