missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylmethanol 97.0+%, TCI America™

Catalog No. p-7136682
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12505ML 5 mL
T125025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12505ML Supplier TCI America Supplier No. T12505ML

Chemical Identifiers

CAS 3219-63-4
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.224
MDL Number MFCD00004596
InChI Key ZQKNBDOVPOZPLY-UHFFFAOYSA-N
Synonym trimethylsilyl methanol, 1-trimethylsilylmethanol, hydroxymethyltrimethylsilane, methanol, trimethylsilyl, silane, hydroxymethyl trimethyl, hydroxymethyl trimethylsilane, trimethylhydroxymethylsilane, methanol, 1-trimethylsilyl, trimethylsilyl-methanol, acmc-209hrr
PubChem CID 76692
IUPAC Name trimethylsilylmethanol
SMILES C[Si](C)(C)CO

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 mL
UN Number 1993
Formula Weight 104.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylmethanol
RTECSNumber PC4100000
TSCA Yes