missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L), TCI America™
Supplier: TCI America T1146100ML
Specifications
| Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L) | |
| Yellow | |
| MFCD00053946 | |
| 2924 | |
| ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
| diazomethyl(trimethyl)silane | |
| 167693 | |
| Liquid |
| 18107-18-1 | |
| C4H10N2Si | |
| 100 mL | |
| trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
| C[Si](C)(C)C=[N+]=[N-] | |
| 114.223 | |
| 114.22 |
Chemical Identifiers
| 18107-18-1 | |
| 114.223 | |
| ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
| 167693 | |
| C[Si](C)(C)C=[N+]=[N-] |
| C4H10N2Si | |
| MFCD00053946 | |
| trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
| diazomethyl(trimethyl)silane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator