missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L), TCI America™

Catalog No. p-7136631
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T114610ML 10 mL
T114625ML 25 mL
T1146100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T114610ML Supplier TCI America Supplier No. T114610ML
Only null left

Chemical Identifiers

CAS 18107-18-1
Molecular Formula C4H10N2Si
Molecular Weight (g/mol) 114.223
MDL Number MFCD00053946
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
PubChem CID 167693
IUPAC Name diazomethyl(trimethyl)silane
SMILES C[Si](C)(C)C=[N+]=[N-]

Specifications

Color Yellow
Quantity 10 mL
UN Number 2924
Formula Weight 114.22
Physical Form Liquid
Chemical Name or Material Trimethylsilyldiazomethane (ca. 10% in Hexane, ca. 0.6mol/L)
TSCA No
Recommended Storage Refrigerator