missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Propionate 95.0+%, TCI America™

Catalog No. T35521ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35521ML 1 mL
T35525ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35521ML Supplier TCI America Supplier No. T35521ML

Chemical Identifiers

CAS 16844-98-7
Molecular Formula C6H14O2Si
Molecular Weight (g/mol) 146.26
MDL Number MFCD01632555
InChI Key QVSRWXFOZLIWJS-UHFFFAOYSA-N
Synonym trimethylsilyl propionate, silanol, trimethyl-, propanoate, silanol, trimethyl-, propionate, propionic acid trimethylsilyl ester, trimethylsilanol propanoate, trimethylsilylpropionate, propanoic acid, tms derivative, propanoic acid, trimethylsilyl ester, propionic acid, trimethylsilyl ester, silanol,1,1,1-trimethyl-, 1-propanoate
PubChem CID 519321
IUPAC Name trimethylsilyl propanoate
SMILES CCC(=O)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 122°C
Quantity 1 mL
UN Number 1993
Formula Weight 146.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Propionate
EINECSNumber (9)-1939
TSCA No