CAS RN 16844-98-7
IUPAC Name:
trimethylsilyl propanoate
Synonyms:
trimethylsilyl propanoate
Molecular Weight (g/mol):
146.26
Molecular Formula:
C6H14O2Si
InChi Key:
QVSRWXFOZLIWJS-UHFFFAOYSA-N
SMILES:
CCC(=O)O[Si](C)(C)C
1
–
1
of
1
results
1
Trimethylsilyl Propionate 95.0+%, TCI America™
CAS: 16844-98-7 Molecular Formula: C6H14O2Si Molecular Weight (g/mol): 146.26 MDL Number: MFCD01632555 InChI Key: QVSRWXFOZLIWJS-UHFFFAOYSA-N Synonym: trimethylsilyl propionate,silanol, trimethyl-, propanoate,silanol, trimethyl-, propionate,propionic acid trimethylsilyl ester,trimethylsilanol propanoate,trimethylsilylpropionate,propanoic acid, tms derivative,propanoic acid, trimethylsilyl ester,propionic acid, trimethylsilyl ester,silanol,1,1,1-trimethyl-, 1-propanoate PubChem CID: 519321 IUPAC Name: trimethylsilyl propanoate SMILES: CCC(=O)O[Si](C)(C)C