missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Isocyanate 95.0+%, TCI America™

Supplier:  TCI America T11065G

 View more versions of this product

Catalog No. T11065G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethylsilyl Isocyanate
Colorless
C4H9NOSi
5 g
trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
C[Si](C)(C)N=C=O
115.21
115.21
Liquid
1118-02-1
90°C
MFCD00001993
2478
NIZHERJWXFHGGU-UHFFFAOYSA-N
isocyanatotrimethylsilane
70696
≥95.0% (GC)

Chemical Identifiers

1118-02-1
115.21
NIZHERJWXFHGGU-UHFFFAOYSA-N
70696
C[Si](C)(C)N=C=O
C4H9NOSi
MFCD00001993
trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
isocyanatotrimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions