missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Isocyanate 95.0+%, TCI America™

Catalog No. p-7136611
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
T11065G 5 g
T110625G 25 g
T1106100G 100 g
3 options

Catalog No. T11065G

Supplier: TCI America T11065G

Only null left

Chemical Identifiers

CAS 1118-02-1
Molecular Formula C4H9NOSi
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001993
InChI Key NIZHERJWXFHGGU-UHFFFAOYSA-N
Synonym trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
PubChem CID 70696
IUPAC Name isocyanatotrimethylsilane
SMILES C[Si](C)(C)N=C=O

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 g
UN Number 2478
Formula Weight 115.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator