missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Iodide (stabilized with Aluminum) 95.0+%, TCI America™

Catalog No. p-7131323
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
I03085G 5 g
I030825G 25 g
2 options

Catalog No. I03085G

Supplier: TCI America I03085G

Only null left

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.094
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Boiling Point 106°C
Quantity 5 g
UN Number 2924
Formula Weight 200.09
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Iodide (stabilized with Aluminum)