missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl Difluoro(fluorosulfonyl)acetate 96.0+%, TCI America™
Supplier: TCI America T302225G
Specifications
| Trimethylsilyl Difluoro(fluorosulfonyl)acetate | |
| Colorless | |
| C5H9F3O4SSi | |
| 25 g | |
| trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr | |
| C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O | |
| 250.26 | |
| 250.26 | |
| Liquid |
| 120801-75-4 | |
| 86°C | |
| MFCD02093343 | |
| 2920 | |
| XHVSCKNABCCCAC-UHFFFAOYSA-N | |
| trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate | |
| 2778022 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 120801-75-4 | |
| 250.26 | |
| XHVSCKNABCCCAC-UHFFFAOYSA-N | |
| 2778022 | |
| C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O |
| C5H9F3O4SSi | |
| MFCD02093343 | |
| trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr | |
| trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate |
Safety and Handling
TSCA : No