accessibility menu, dialog, popup

UserName

CH3SiCH3H3COOFFSOOF

CAS RN 120801-75-4

IUPAC Name: trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Synonyms: trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Molecular Weight (g/mol): 250.26
Molecular Formula: C5H9F3O4SSi
InChi Key: XHVSCKNABCCCAC-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)