missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Difluoro(fluorosulfonyl)acetate 96.0+%, TCI America™

Catalog No. T30225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30225G 5 g
T302225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30225G Supplier TCI America Supplier No. T30225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120801-75-4
Molecular Formula C5H9F3O4SSi
Molecular Weight (g/mol) 250.26
MDL Number MFCD02093343
InChI Key XHVSCKNABCCCAC-UHFFFAOYSA-N
Synonym trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr
PubChem CID 2778022
IUPAC Name trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O

Specifications

Color Colorless
Boiling Point 86°C
Quantity 5 g
UN Number 2920
Formula Weight 250.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Difluoro(fluorosulfonyl)acetate