missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes

Catalog No. AC385331000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC385331000 100 mL Glass bottle
AC385330050 5 mL Glass bottle
AC385330250 25 mL Glass bottle
AC385335000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC385331000 Supplier Thermo Scientific Chemicals Supplier No. 385331000
Only null left

CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 18107-18-1 , 92112-69-1
Molecular Formula C4H10N2Si
Molecular Weight (g/mol) 114.22
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
PubChem CID 167693
IUPAC Name diazomethyl(trimethyl)silane
SMILES C[Si](C)(C)C=[N+]=[N-]

Specifications

CAS Min % 32.15, 64.47
CAS Max % 35.54, 67.85
Color Yellow
Density 0.6750g/mL
Boiling Point 96.0°C
Flash Point −23°C
Packaging Glass bottle
Linear Formula (CH3)3SiCHN2
Quantity 100 mL
Specific Gravity 0.675
Solubility Information Solubility in water: insoluble
Formula Weight 114.22
Grade Technical
Physical Form Solution
Chemical Name or Material (Trimethylsilyl)diazomethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes skin irritation.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Obtain special instructions before use.
Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot su

RUO – Research Use Only