CAS RN 92112-69-1
Hexanes (Certified ACS), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (HPLC), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (Spectranalyzed™), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (GC Resolv™), Fisher Chemical
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (Certified), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: Naptha Solvent,Petroleum Naptha Solvent PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (Pesticide), Fisher Chemical
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, 99%, pure, mixture of isomers
CAS: 92112-69-1 | C6H14
Hexanes, mixed isomers, (60+% n-hexane)
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, for HPLC
CAS: 92112-69-1 | C6H14
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
CAS: 92112-69-1 | C6H14
Hexanes, technical, mixture of isomers
CAS: 92112-69-1 | C6H14
Hexanes, 99%, for spectroscopy ACS, mixture of isomers
CAS: 92112-69-1 | C6H14
Hexanes, for analysis, mixture of isomers
CAS: 92112-69-1 | C6H14
Hexanes, 99%, ACS reagent, mixture of isomers
CAS: 92112-69-1 Molecular Formula: C6H14 MDL Number: MFCD00009520 Synonym: Hex
- 1
- 2