missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Specifications
| CAS Min % | 32.15, 64.47 |
| CAS Max % | 35.54, 67.85 |
| Color | Yellow |
| Density | 0.6750g/mL |
| Boiling Point | 96.0°C |
| Flash Point | −23°C |
| Packaging | Glass bottle |
| Linear Formula | (CH3)3SiCHN2 |
| Quantity | 5 mL |
| Specific Gravity | 0.675 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement Fatal if inhaled. Causes skin irritation. Suspected of damaging fertility. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged |
| missing translation for 'healthHazard3' | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Obtain special instructions before use. Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot su |
RUO – Research Use Only