missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl Acrylate (stabilized with BHT) 95.0+%, TCI America™
Supplier: TCI America T355925ML
Specifications
| Trimethylsilyl Acrylate (stabilized with BHT) | |
| Yellow | |
| C6H12O2Si | |
| 25 mL | |
| Acrylic Acid Trimethylsilyl Ester | |
| C[Si](C)(C)OC(=O)C=C | |
| 144.245 | |
| 144.25 | |
| Liquid |
| 13688-55-6 | |
| 54°C | |
| MFCD00156432 | |
| 1993 | |
| OTYBJBJYBGWBHB-UHFFFAOYSA-N | |
| trimethylsilyl prop-2-enoate | |
| 554662 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 13688-55-6 | |
| 144.245 | |
| OTYBJBJYBGWBHB-UHFFFAOYSA-N | |
| 554662 | |
| C[Si](C)(C)OC(=O)C=C |
| C6H12O2Si | |
| MFCD00156432 | |
| Acrylic Acid Trimethylsilyl Ester | |
| trimethylsilyl prop-2-enoate |
Safety and Handling
TSCA : No