missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Acrylate (stabilized with BHT) 95.0+%, TCI America™

Catalog No. p-7137798
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35595ML 5 mL
T355925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35595ML Supplier TCI America Supplier No. T35595ML

Chemical Identifiers

CAS 13688-55-6
Molecular Formula C6H12O2Si
Molecular Weight (g/mol) 144.245
MDL Number MFCD00156432
InChI Key OTYBJBJYBGWBHB-UHFFFAOYSA-N
Synonym Acrylic Acid Trimethylsilyl Ester
PubChem CID 554662
IUPAC Name trimethylsilyl prop-2-enoate
SMILES C[Si](C)(C)OC(=O)C=C

Specifications

Color Yellow
Boiling Point 54°C
Quantity 5 mL
UN Number 1993
Formula Weight 144.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Acrylate (stabilized with BHT)