missing translation for 'onlineSavingsMsg'
Learn More

Trimethylpropylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7137400
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27615G 5 g
T276125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27615G Supplier TCI America Supplier No. T27615G

Chemical Identifiers

CAS 268536-05-6
Molecular Formula C8H16F6N2O4S2
Molecular Weight (g/mol) 382.336
MDL Number MFCD08458920
InChI Key NFLGAVZONHCOQE-UHFFFAOYSA-N
PubChem CID 53384436
IUPAC Name bis(trifluoromethylsulfonyl)azanide;trimethyl(propyl)azanium
SMILES CCC[N+](C)(C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 19°C
Color Yellow
Quantity 5 g
Formula Weight 382.34
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethylpropylammonium Bis(trifluoromethanesulfonyl)imide