accessibility menu, dialog, popup

UserName

CH3NCH3CH3H3CFFFSOONSOOFFF

CAS RN 268536-05-6

IUPAC Name: trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane; trimethyl(propyl)azanium
Synonyms: bistriflylimide anion; nervonyl carnitine
Molecular Weight (g/mol): 382.34
Molecular Formula: C8H16F6N2O4S2
InChi Key: NFLGAVZONHCOQE-UHFFFAOYSA-N
SMILES: CCC[N+](C)(C)C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results