missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Bromide 98.0+%, TCI America™

Catalog No. P024325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024325G 25 g
P0243100G 100 g
P0243500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P024325G Supplier TCI America Supplier No. P024325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16056-11-4
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.122
MDL Number MFCD00011788
InChI Key GNMJFQWRASXXMS-UHFFFAOYSA-M
Synonym trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide
PubChem CID 27663
IUPAC Name trimethyl(phenyl)azanium;bromide
SMILES C[N+](C)(C)C1=CC=CC=C1.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 216.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylphenylammonium Bromide
EINECSNumber (9)-1187
RTECSNumber BT2100000
TSCA Yes