missing translation for 'onlineSavingsMsg'
Learn More

Trimethyloxonium Tetrafluoroborate 95.0+%, TCI America™

Catalog No. T15075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15075G 5 g
T150725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15075G Supplier TCI America Supplier No. T15075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 420-37-1
Molecular Formula C3H9BF4O
Molecular Weight (g/mol) 147.91
MDL Number MFCD00011798
InChI Key CZVZBKHWOFJNCR-UHFFFAOYSA-N
Synonym trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate
PubChem CID 2735153
IUPAC Name tetrafluoroboranuide; trimethyloxidanium
SMILES C[O+](C)C.F[B-](F)(F)F

Specifications

Quantity 5 g
UN Number 1759
Formula Weight 147.91
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyloxonium Tetrafluoroborate
TSCA Yes
Recommended Storage Freezer