accessibility menu, dialog, popup

UserName

CAS RN 420-37-1

IUPAC Name: tetrafluoroboranuide; trimethyloxidanium
Synonyms: trimethyloxonium tetrafluoroborate
Molecular Weight (g/mol): 147.91
Molecular Formula: C3H9BF4O
InChi Key: CZVZBKHWOFJNCR-UHFFFAOYSA-N
SMILES: C[O+](C)C.F[B-](F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)