missing translation for 'onlineSavingsMsg'
Learn More

Trimethyloxonium tetrafluoroborate, 95%

Catalog No. AC421960025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421960025 2.5 g Glass bottle
AC421960100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421960025 Supplier Thermo Scientific Chemicals Supplier No. 421960025

CAS: 420-37-1 | C3H9BF4O | 147.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 420-37-1
Molecular Formula C3H9BF4O
Molecular Weight (g/mol) 147.91
MDL Number MFCD00011798
InChI Key CZVZBKHWOFJNCR-UHFFFAOYSA-N
Synonym trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate
PubChem CID 2735153
IUPAC Name trimethyloxidanium;tetrafluoroborate
SMILES C[O+](C)C.F[B-](F)(F)F

Specifications

Melting Point 200°C
Color White
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)3OBF4
Quantity 2.5 g
UN Number 3261
Fieser 01,1232; 03,438; 04,314; 05,541; 13,327
Merck Index 15, 9889
Formula Weight 147.91
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material Trimethyloxonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 206-994-4

RUO – Research Use Only