missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane Tris(thioglycolate) 80.0+%, TCI America™

Catalog No. p-7137328
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T263325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T263325G Supplier TCI America Supplier No. T263325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10193-96-1
Molecular Formula C12H20O6S3
Molecular Weight (g/mol) 356.466
MDL Number MFCD00046836
InChI Key KAJBSGLXSREIHP-UHFFFAOYSA-N
Synonym 1,1,1-Tris(thioglycoloyloxymethyl)propane
PubChem CID 82439
IUPAC Name 2,2-bis[(2-sulfanylacetyl)oxymethyl]butyl 2-sulfanylacetate
SMILES CCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 g
Formula Weight 356.47
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylolpropane Tris(thioglycolate)
EINECSNumber (7)-1074
TSCA Yes