accessibility menu, dialog, popup

UserName

CAS RN 10193-96-1

IUPAC Name: 2,2-bis({[(2-sulfanylacetyl)oxy]methyl})butyl 2-sulfanylacetate
Synonyms: 2,2-bis({[(2-sulfanylacetyl)oxy]methyl})butyl 2-sulfanylacetate
Molecular Weight (g/mol): 356.47
Molecular Formula: C12H20O6S3
InChi Key: KAJBSGLXSREIHP-UHFFFAOYSA-N
SMILES: CCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results