missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane Tris(3-mercaptopropionate) 85.0+%, TCI America™

Catalog No. p-7137321
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T261925G 25 g
T2619500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T261925G Supplier TCI America Supplier No. T261925G

Chemical Identifiers

CAS 33007-83-9
Molecular Formula C15H26O6S3
Molecular Weight (g/mol) 398.547
MDL Number MFCD00046842
InChI Key IMQFZQVZKBIPCQ-UHFFFAOYSA-N
Synonym 1,1,1-Tris(3-mercaptopropionyloxymethyl)propane
PubChem CID 118379
IUPAC Name 2,2-bis(3-sulfanylpropanoyloxymethyl)butyl 3-sulfanylpropanoate
SMILES CCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 398.55
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylolpropane Tris(3-mercaptopropionate)